Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGOL2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB291637 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291637 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291637 Supplier Novus Biologicals Supplier No. NB10060454PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGOL2 Polyclonal antibody specifically detects SGOL2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen SGOL2
Applications Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias FLJ25211, MGC129665, Sgo2, Shugoshin-2, shugoshin-like 2 (S. pombe), Tripin, TRIPINshugoshin-like 2
Host Species Rabbit
Immunogen Maps to a region between residue 175 and 225 of human shugoshin-like 2 using the numbering given in entry NP_689737.3 (GeneID 151246).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151246
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.