Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGPP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
175 μg
25 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151764 25 μg
NB523807 175 μg
NB151763 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB151764 Supplier Novus Biologicals Supplier No. NBP31190525UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGPP1 Polyclonal antibody specifically detects SGPP1 in Human samples. It is validated for Western Blot

Specifications

Antigen SGPP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:200
Formulation PBS
Gene Alias sphingosine-1-phosphate phosphatase 1
Host Species Rabbit
Immunogen Synthetic peptide derived from the SGPP1 protein. (Uniprot: Q9BX95)
Purification Method Ammonium sulfate precipitation
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81537
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.