Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGSM2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341535 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341535 Supplier Novus Biologicals Supplier No. NBP282004MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SGSM2 Polyclonal antibody specifically detects SGSM2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SGSM2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias KIAA0397, RUN and TBC1 domain containing 1, RUN and TBC1 domain-containing protein 1, RUTBC1, small G protein signaling modulator 2, small G protein signaling modulator 2 protein
Host Species Rabbit
Immunogen SGSM2 antibody was raised against an 18 amino acid synthetic peptide near the center of human SGSM2. The immunogen is located within amino acids 650 - 700 of SGSM2. Amino Acid Squence: VDDLEPPEPQDPEDSRPK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9905
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.