Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SGSM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519154
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519154 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519154 Supplier Invitrogen™ Supplier No. PA519154
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

May play a cooperative role in NF2-mediated growth suppression of cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Sgsm3
Gene Accession No. Q96HU1
Gene Alias 1810012I01Rik; AI428557; BB175482; Cip85; Cx43-interacting protein of 85-kDa; DKFZp761D051; MAP; merlin binding protein; merlin-associated protein; R75178; RABGAP5; RabGAP-5; RabGAPLP; rab-GTPase-activating protein-like protein; RUN and SH3 containing 3; RUN and TBC1 domain containing 3; RUN and TBC1 domain-containing protein 3; RUSC3; Rutbc3; SGSM3; small G protein signaling modulator 3; small G protein signaling modulator 3 protein
Gene Symbols Sgsm3
Host Species Goat
Immunogen Synthetic peptide sequence (RDHENYVACSRSHRR) corresponding to the internal amino acids of SGSM3 (aa 469-484).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27352
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.