Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SGTA Antibody (6A4), Novus Biologicals™
SDP

Catalog No. NB177216 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177216 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177216 Supplier Novus Biologicals Supplier No. NBP313529
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

SGTA Monoclonal antibody specifically detects SGTA in Human, Monkey samples. It is validated for Western Blot

Specifications

Antigen SGTA
Applications Western Blot
Classification Monoclonal
Clone 6A4
Conjugate Unconjugated
Formulation PBS
Gene Alias alphaSGT, Alpha-SGT, protein containing three tetratricopeptide repeats, SGT1, SGThSGT, small glutamine-rich tetratricopeptide repeat (TPR)-containing, small glutamine-rich tetratricopeptide repeat (TPR)-containing, alpha, small glutamine-rich tetratricopeptide repeat-containing protein alpha, UBP, Vpu-binding protein
Host Species Mouse
Immunogen The immunogen used to generate this antibody corresponds to human SGTA.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6449
Target Species Human, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.