Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2D1A Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB089210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089210 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089210 Supplier Novus Biologicals Supplier No. NBP299726AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SH2D1A Polyclonal specifically detects SH2D1A in Human samples. It is validated for ELISA.

Specifications

Antigen SH2D1A
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant SH2D1A (NP_002342.1; Pro2-Lys97).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4068
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.