Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2D2A Antibody (OTI4F3), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP27413A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27413A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27413A32 Supplier Novus Biologicals Supplier No. NBP274153AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH2D2A Monoclonal specifically detects SH2D2A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SH2D2A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4F3
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias F2771, SCAP, SH2 domain containing 2A, SH2 domain protein 2A, SH2 domain-containing adapter protein, T cell specific adapter protein TSAd, T cell-specific adapter protein, T lymphocyte specific adaptor protein, TSAd, TSADSH2 domain-containing protein 2A, VEGF receptor-associated protein, VRAP
Gene Symbols SH2D2A
Host Species Mouse
Immunogen Full length recombinant protein of human SH2D2A (NP_003966) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9047
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.