Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH2D2A Antibody (OTI4F3), Novus Biologicals™
SDP

Catalog No. NBP201828 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP201828 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP201828 Supplier Novus Biologicals Supplier No. NBP201828
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH2D2A Monoclonal specifically detects SH2D2A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SH2D2A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4F3
Concentration 0.71 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150
Gene Alias F2771, SCAP, SH2 domain containing 2A, SH2 domain protein 2A, SH2 domain-containing adapter protein, T cell specific adapter protein TSAd, T cell-specific adapter protein, T lymphocyte specific adaptor protein, TSAd, TSADSH2 domain-containing protein 2A, VEGF receptor-associated protein, VRAP
Gene Symbols SH2D2A
Host Species Mouse
Immunogen Full length human recombinant protein of human SH2D2A(NP_003966) produced in HEK293T cell.
Molecular Weight of Antigen 42.8 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 9047
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.