Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7206106 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-9041 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-9041 Supplier Proteintech Group Inc Supplier No. 138491AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SH3GL3 is implicated in endocytosis. It may recruit other proteins to membranes with high curvature.

Specifications

Antigen SH3GL3
Applications Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SH3GL3
Gene Accession No. O35180, Q62421, Q99963
Gene Alias CNSA3, EEN 2B L3, EEN B2, Endophilin 3, Endophilin A3, HsT19371, SH3 domain GRB2 like 3, SH3 domain protein 2C, SH3D2C, SH3GL3, SH3P13
Gene Symbols SH3GL3
Host Species Rabbit
Immunogen SH3GL3 Fusion Protein Ag4819
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20408, 6457, 81921
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.