Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3GLB2 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB331657 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB331657 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB331657 Supplier Novus Biologicals Supplier No. NBP297456APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SH3GLB2 Polyclonal antibody specifically detects SH3GLB2 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SH3GLB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias endophilin-B2, GRB2-like, endophilin B2, SH3-containing protein SH3GLB2, SH3-domain GRB2-like endophilin B2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SH3GLB2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 56904
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.