Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3MD2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP154972 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP154972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP154972 Supplier Novus Biologicals Supplier No. NBP154972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SH3MD2 Polyclonal specifically detects SH3MD2 in Human samples. It is validated for Western Blot.

Specifications

Antigen SH3MD2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q7Z6J0
Gene Alias EC 6.3.2, EC 6.3.2.-, KIAA1494SH3 multiple domains protein 2, Plenty of SH3s, POSHFLJ21602, Protein POSH, putative E3 ubiquitin-protein ligase SH3RF1, RING finger protein 142, RNF142plenty of SH3 domains, SH3 domain containing ring finger 1, SH3 domain-containing RING finger protein 1, SH3 multiple domains 2, SH3MD2
Gene Symbols SH3RF1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SH3RF1(SH3 domain containing ring finger 1) The peptide sequence was selected from the middle region of SH3RF1. Peptide sequence LLKLLSGASTKRKPRVSPPASPTLEVELGSAELPLQGAVGPELPPGGGHG.
Molecular Weight of Antigen 93 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 57630
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.