Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3PX1 Antibody (OTI1B5), FITC, Novus Biologicals™
SDP

Catalog No. NBP274158F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274158F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274158F Supplier Novus Biologicals Supplier No. NBP274158F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH3PX1 Monoclonal specifically detects SH3PX1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SH3PX1
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B5
Conjugate FITC
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias MST155, MSTP155, Protein SDP1, SDP1, SH3 and PX domain-containing protein 1, SH3PX1WISP, SH3PXD3ASH3 and PX domain-containing protein SH3PX1, sorting nexin 9, sorting nexin-9, WiskSH3 and PX domain-containing protein 3A
Gene Symbols SNX9
Host Species Mouse
Immunogen Full length recombinant protein of human SH3PX1 (NP_057308) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51429.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.