Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SH3PX1 Antibody (OTI1B5), Novus Biologicals™
SDP

Catalog No. NBP202609 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP202609 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP202609 Supplier Novus Biologicals Supplier No. NBP202609
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SH3PX1 Monoclonal specifically detects SH3PX1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SH3PX1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI1B5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50
Gene Alias MST155, MSTP155, Protein SDP1, SDP1, SH3 and PX domain-containing protein 1, SH3PX1WISP, SH3PXD3ASH3 and PX domain-containing protein SH3PX1, sorting nexin 9, sorting nexin-9, WiskSH3 and PX domain-containing protein 3A
Gene Symbols SNX9
Host Species Mouse
Immunogen Full length human recombinant protein of human SNX9(NP_057308) produced in HEK293T cell.
Molecular Weight of Antigen 66.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51429
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.