Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHANK1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications
$243.50 - $499.50
Specifications
Antigen | SHANK1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:250-1:750, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin 1:250-1:750 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry |
Classification | Polyclonal |
Description
SHANK1 Polyclonal specifically detects SHANK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SHANK1 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:250-1:750, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin 1:250-1:750 | |
Polyclonal | |
Rabbit | |
Neuroscience | |
PBS with 0.02% Sodium Azide | |
50944 | |
residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon | |
SHANK1 | |
IgG | |
Affinity Purified | |
Reacts with residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title