Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018146 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018146 0.025 mL
NB300167 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018146 Supplier Novus Biologicals Supplier No. NB300167SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 6 publications

SHANK1 Polyclonal specifically detects SHANK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SHANK1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:250-1:750, Immunocytochemistry/Immunofluorescence 1:800, Immunohistochemistry-Paraffin 1:250-1:750
Formulation PBS with 0.02% Sodium Azide
Gene Alias SH3 and multiple ankyrin repeat domains 1, SH3 and multiple ankyrin repeat domains protein 1, Shank1, Somatostatin receptor-interacting protein, SSTR-interacting protein, SSTRIPSPANK-1, synamon
Gene Symbols SHANK1
Host Species Rabbit
Immunogen residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 50944
Test Specificity Reacts with residues [SGPIYPGLFDIRSS] of the C-terminus of the SHANK1 protein.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.