Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK3 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343869 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343869 Supplier Novus Biologicals Supplier No. NBP241171MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHANK3 Polyclonal antibody specifically detects SHANK3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SHANK3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the center of human SHANK3. The immunogen is located within amino acids 890 - 940 of SHANK3. Amino Acid Squence: APRPPPAATPPERPKRRPR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 85358
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.