Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK3 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB289906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289906 Supplier Novus Biologicals Supplier No. NBP232161PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHANK3 Polyclonal antibody specifically detects SHANK3 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen SHANK3
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1475 to 1525 of human SH3 and Multiple Ankyrin Repeat Domains 3 using the numbering given in entry Q9BYB0
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 85358
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.