Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHANK3 Antibody (S367-51), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB242189488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB242189488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB242189488 Supplier Novus Biologicals Supplier No. NBP242189AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHANK3 Monoclonal specifically detects SHANK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SHANK3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S367-51
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DEL22q13.3, KIAA1650proline rich synapse associated protein 2, Proline-rich synapse-associated protein 2, ProSAP2, PSAP2SH3 and multiple ankyrin repeat domains protein 3, SH3 and multiple ankyrin repeat domains 3, shank postsynaptic density protein, Shank3, SPANK-2
Gene Symbols SHANK3
Host Species Mouse
Immunogen Fusion protein amino acids 538-626 (SH3 domain) of rat SHANK3. Mouse: 100% identity (89/89 amino acids identical). Human: 97% identity (87/89 amino acids identical). ∼70% identity with SHANK1 and SHANK2.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 85358
Test Specificity Approx 70% identity with SHANK1 and SHANK2.Detects approx 190kDa. Cross-reacts with SHANK1. Does not cross-react with SHANK2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.