Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHC1 Antibody (6E10), Novus Biologicals™
SDP

Catalog No. NB036642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036642 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036642 Supplier Novus Biologicals Supplier No. NBP289051
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHC1 Monoclonal specifically detects SHC1 in Human, Mouse, Canine samples. It is validated for Western Blot.

Specifications

Antigen SHC1
Applications Western Blot
Classification Monoclonal
Clone 60000000000
Conjugate Unconjugated
Formulation Lyophilized from 1ml of 2x PBS, PEG, and sucrose
Gene Alias FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1
Host Species Mouse
Immunogen Synthetic peptide corresponding to aa 33-39 (E33LPpSPSA39) of SHC1 phosphorylated at Ser36 conjugated to KLH.
Purification Method Multi-step
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6464
Reconstitution Reconstitute with 1 mL water (15 minutes at room temperature).
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.