Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHC1 Antibody (SR1785), Novus Biologicals™
SDP

Catalog No. NB387941 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387941 100 μg
NB387940 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387941 Supplier Novus Biologicals Supplier No. NBP321850100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SHC1 Monoclonal antibody specifically detects SHC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SHC1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1785
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200
Formulation PBS (pH 7.4), 150 mM NaCl, 50% Glycerol
Gene Alias FLJ26504, p66, p66SHC, SH2 domain protein C1, SHC, SHC (Src homology 2 domain containing) transforming protein 1, SHC (Src homology 2 domain-containing) transforming protein 1, SHCA, SHC-transforming protein 1, SHC-transforming protein 3, SHC-transforming protein A, Src homology 2 domain-containing-transforming protein C1
Host Species Rabbit
Immunogen A synthesized peptide derived from human SHC1 (Uniprot #: P29353)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6464
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.