Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigatoxin 1 Antibody (13C4), Novus Biologicals™
SDP

Catalog No. NB119875 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119875 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119875 Supplier Novus Biologicals Supplier No. NBP311250
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Shigatoxin 1 Monoclonal specifically detects Shigatoxin 1 in Bacteria samples. It is validated for Western Blot, Functional, Immunoassay.

Specifications

Antigen Shigatoxin 1
Applications Western Blot, Functional Assay, Immunoassay
Classification Monoclonal
Clone 13C4
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Functional, Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Shiga toxin 1 subunit B, Shiga-like toxin 1, Stx1, STX1B, Verotoxin 1, VT-1
Host Species Mouse
Immunogen The monoclonal antibody 13C4 recognizes the 1B subunit of Shigatoxin 1.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 916677
Target Species Bacteria
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.