Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Shigella Antibody (306/305) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | Shigella |
|---|---|
| Clone | 306/305 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017518
![]() |
Novus Biologicals
NB100629180.025MG |
0.025 mg |
Each for $181.90
|
|
|||||
NB10062918
![]() |
Novus Biologicals
NB10062918 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Shigella Monoclonal specifically detects Shigella in Bacteria samples. It is validated for ELISA.Specifications
| Shigella | |
| 1.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| RUO | |
| Shigella Boydii; Shigella Dysenteriae; Shigella Flexneri; Shigella Sonnei | |
| IgG1 | |
| Affinity Purified | |
| This product is specific for Shigella flexneri serotype 6. S. flexneri is a gram negative, rod shaped bacteria that is very closely related to E. coli and Salmonella. |
| 306/305 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Bacteria | |
| NCTC 9729 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title