Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella Antibody (306/305), HRP, Novus Biologicals™
SDP

Catalog No. NB10062918H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062918H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062918H Supplier Novus Biologicals Supplier No. NB10062918H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Shigella Monoclonal specifically detects Shigella in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Shigella
Applications ELISA
Classification Monoclonal
Clone 306/305
Conjugate HRP
Dilution ELISA
Formulation PBS with No Preservative
Gene Alias Shigella Boydii; Shigella Dysenteriae; Shigella Flexneri; Shigella Sonnei
Host Species Mouse
Immunogen NCTC 9729
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product is specific for Shigella flexneri serotype 6. S. flexneri is a gram negative, rod shaped bacteria that is very closely related to E. coli and Salmonella.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.