Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella Dysenteriae Serotype 1 Antibody (35/383), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N106468A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106468A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106468A70 Supplier Novus Biologicals Supplier No. NB10064568AF750

Mouse Monoclonal Antibody

Shigella Dysenteriae Serotype 1 Monoclonal specifically detects Shigella Dysenteriae Serotype 1 in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Shigella Dysenteriae Serotype 1
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 35/383
Conjugate Alexa Fluor 750
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias S. boydii, S. dysenteriae, S. flexneri, S. sonnei, Shigella Boydii, Shigella Dysenteriae, Shigella Flexneri, Shigella Sonnei
Host Species Mouse
Immunogen Native S. dysenteriae, NCTC 4837 (ATCC 13313).
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Reacts with NCTC 4837. Shigella Dysenteriae Serotype 1
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.