Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella Dysenteriae Serotype 1 Antibody (35/383), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB10064568G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064568G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10064568G Supplier Novus Biologicals Supplier No. NB10064568G

Mouse Monoclonal Antibody

Shigella Dysenteriae Serotype 1 Monoclonal specifically detects Shigella Dysenteriae Serotype 1 in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Shigella Dysenteriae Serotype 1
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 35/383
Conjugate DyLight 488
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias S. boydii, S. dysenteriae, S. flexneri, S. sonnei, Shigella Boydii, Shigella Dysenteriae, Shigella Flexneri, Shigella Sonnei
Host Species Mouse
Immunogen Native S. dysenteriae, NCTC 4837 (ATCC 13313).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with NCTC 4837. Shigella Dysenteriae Serotype 1
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.