Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shigella Group Antigen Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017691 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017691 0.1 mL
NB10065058 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017691 Supplier Novus Biologicals Supplier No. NB100650580.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Shigella Group Antigen Polyclonal specifically detects Shigella Group Antigen in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Shigella Group Antigen
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Gene Alias S. boydii, S. dysenteriae, S. flexneri, S. sonnei, Shigella Boydii, Shigella Dysenteriae, Shigella Flexneri, Shigella Sonnei
Host Species Rabbit
Immunogen Mixture of S. boydii, S. flexneri and S. dysenteriae.
Purification Method IgG purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65058 is polyvalent for Shigella antigens. Shigella sp. are non spore forming, rod shaped bacteria, the causitive agent of Shigellosis in man. The genus is closely related to Escherichia coli and Salmonella . The antibody preparation has not been cross absorbed and is likely to react with closely related Enterobacteriaceae
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.