Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Shigella Group Antigen Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Shigella Group Antigen |
---|---|
Concentration | 5.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry, Immunohistochemistry, ELISA, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017691
![]() |
Novus Biologicals
NB100650580.1ML |
0.1 mL |
Each for $206.50
|
|
|||||
NB10065058
![]() |
Novus Biologicals
NB10065058 |
0.5 mL |
Each for $560.50
|
|
|||||
Description
Shigella Group Antigen Polyclonal specifically detects Shigella Group Antigen in Bacteria samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
Shigella Group Antigen | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline with 0.09% Sodium Azide | |
Mixture of S. boydii, S. flexneri and S. dysenteriae. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
5.0 mg/mL | |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry, Immunohistochemistry, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Bacteria | |
S. boydii, S. dysenteriae, S. flexneri, S. sonnei, Shigella Boydii, Shigella Dysenteriae, Shigella Flexneri, Shigella Sonnei | |
IgG | |
IgG purified | |
NB100-65058 is polyvalent for Shigella antigens. Shigella sp. are non spore forming, rod shaped bacteria, the causitive agent of Shigellosis in man. The genus is closely related to Escherichia coli and Salmonella . The antibody preparation has not been cross absorbed and is likely to react with closely related Enterobacteriaceae |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title