Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHIP Antibody (SHIP-01), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB181538549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB181538549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB181538549 Supplier Novus Biologicals Supplier No. NB11081538JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHIP Monoclonal specifically detects SHIP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SHIP
Applications Flow Cytometry
Classification Monoclonal
Clone SHIP-01
Conjugate Janelia Fluor 549
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140
Gene Symbols INPP5D
Host Species Mouse
Immunogen Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3635
Test Specificity The SHIP-01 reacts with SHIP-1, a phosphoinositide phosphatase largely confined to hematopoietic cells. Multiple forms of SHIP-1 have been reported with molecular weights of 110, 125, 130, 135 and 145 kDa.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.