Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHIP Antibody (SHIP-02), FITC, Novus Biologicals™
SDP

Catalog No. NB358782 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358782 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358782 Supplier Novus Biologicals Supplier No. NBP145077F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHIP Monoclonal antibody specifically detects SHIP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular), CyTOF

Specifications

Antigen SHIP
Applications Western Blot, Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone SHIP-02
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Flow (Intracellular), CyTOF-ready
Formulation PBS
Gene Alias 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, MGC104855, SHIPMGC142140
Host Species Mouse
Immunogen Peptide coresponding to a sequence within N-terminal domain of Human SHIP-1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3635
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.