Learn More
Invitrogen™ SHIP1 Recombinant Rabbit Monoclonal Antibody (2F8U0)

Description
Positive test controls include: Raji, U-937. The target is usually found in the following locations: Cytoplasm, Membrane, Peripheral membrane protein. Immunogen sequence: RSFTCSSSAE GRAAGGDKSQ GKPKTPVSSQ APVPAKRPIK PSRSEINQQT PPTPTPRPPL PVKSPAVLHL QHSKGRDYRD NTELPHHGKH RPEEG.
Specifications
Specifications
| Antigen | SHIP1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 2F8U0 |
| Concentration | 0.75 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | INPP5D |
| Gene Accession No. | P97573, Q92835, Q9ES52 |
| Gene Alias | 7a33; hp51CN; Inositol polyphosphate-5-phosphatase 145 kDa; inositol polyphosphate-5-phosphatase D; Inositol polyphosphate-5-phosphatase of 145 kDa; inositol polyphosphate-5-phosphatase, 145 kDa; inositol polyphosphate-5-phosphatase, 145kD; inositol polyphosphate-5-phosphatase, 145kDa; Inpp5d; MGC104855; MGC142140; MGC142142; OTTHUMP00000165069; OTTHUMP00000165070; OTTHUMP00000165071; OTTHUMP00000165072; OTTHUMP00000203442; p150Ship; phosphatidylin; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 1; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 1; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; SH2 domain-containing inositol 5'-phosphatase 1; SH2 domain-containing inositol phosphatase 1; SH2 domain-containing inositol-5'-phosphatase 1; SH2-containing inositol phosphatase SHIP; SHIP; SHIP1; SHIP-1; signaling inositol polyphosphate 5 phosphatase SIP-145; signaling inositol polyphosphate phosphatase SHIP II; SIP-145; Src homology 2 domain-containing inositol-5-phosphatase; s-SHIP |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.