Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHIP2/INPPL1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292556 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292556 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292556 Supplier Novus Biologicals Supplier No. NB1001592APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHIP2/INPPL1 Polyclonal specifically detects SHIP2/INPPL1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen SHIP2/INPPL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 3.1.3, EC 3.1.3.n1, inositol polyphosphate phosphatase-like 1, Inositol polyphosphate phosphatase-like protein 1, INPPL-1, phosphatidylinositol-34,5-trisphosphate 5-phosphatase 2, Protein 51C, SH2 domain-containing inositol phosphatase 2, SH2 domain-containing inositol-5'-phosphatase 2, SHIP-2, SHIP251C protein
Host Species Rabbit
Immunogen Maps to a region between residues 100 and 150 of human inositol polyphosphate phosphatase-like 1 using the numbering given in entry NP_001558.2 (GeneID 3636).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immune System Diseases, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3636
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.