Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 460 Series PVC-Coated Cold Resistant Gloves

Catalog No. 19270239 Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Insulated gloves protect hands, and remain flexible in low temperatures. Rough-textured PVC coating protects against oils and chemicals, while also enhancing grip capabilities and tactile precision.

  • Gloves remain flexible in temperatures as low as -4°F (-20°C)
  • PVC coating protects against oils and chemicals
  • Rough grip enhances tactile precision when handling small, greasy components
  • Extended wrist protection from harmful substances
  • Reduced potential exposure to bacteria, viruses, and fungi

Specifications

Coated Area Full
Coating Material PVC
Color Orange
Exterior Finish Fully Textured
Hand Specific
Size Large
For Use With (Application) Chemical Spray/Treatment, Coating Preparation, and Painting
Glove Cuff Style Gauntlet
Liner Material Acrylic
Series 460
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.