Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Polyurethane Coated Seamless Knit Nylon Gloves

Catalog No. 191304682B Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Catalog No. 19-130-4682B

Supplier: SHOWA™ BO500WM

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves are designed for precision and cleanliness with a permeable nitrile coating, breathable back, and seamless liner to prevent irritation.

  • Snug fit maximizes tactile feedback
  • Coating is permeable, allowing hands to breathe to reduce perspiration
  • Light, elastic low-lint glove is resistant to deformation
  • Polyurethane protects the hand from oils and abrasion
  • Seamless construction eliminates irritation
  • White color is ideal for clean room applications
  • Mimimizes particulates
  • No latex allergy risks

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Cuff Material Elastic
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
EN388 Standard 3131X
For Use With (Application) Intricate Parts Handling, Clean Rooms, Automotive Repairs, and Integrated Circuits
Glove Cuff Color Natural
Glove Cuff Style Knit
Series BO500W
Yarn Gauge 13
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.