Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ 7166 Nitrile Coated Oil & Grease Resistant Gloves

Catalog No. 1138829 Shop All Showa Best Glove Products
Change view
Click to view available options
Exterior Finish:
Fully Textured
No Texture
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Exterior Finish
11-388-29 No Texture
11-394-116 Fully Textured
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 11-388-29 Supplier SHOWA™ Supplier No. 716610
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Full nitrile coating is impermeable to oils and chemicals, while also protecting against abrasions and punctures. Soft cotton liner is soft, comfortable, wicks away sweat, and controls temperature.

  • Exclusive coating provides cut, puncture, snag, and abrasion resistance
  • Smooth finish enhances grip in dry and oily conditions
  • Rough coating enhances grip and adds durability
  • Barrier is resistant to oil and grease, and prevents oil breakthrough
  • Reinforced safety cuff for additional protection
  • Economical replacement for leather

Specifications

Construction Cut and Sewn
Cuff Material Reinforced Cotton
Coating Material Nitrile
Color Black
Exterior Finish No Texture
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size Large
For Use With (Application) Chemical Spray/Treatment, Plumbing, Landscaping, and Utility Work
Glove Cuff Color Natural
Glove Cuff Style Safety
Packaging Type Polybag
Series 7166
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.