Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ Zorb-IT™ Blacklite Nitrile Foam Coated Nylon Gloves

Catalog No. 19172195 Shop All Showa Best Glove Products
Click to view available options
Size:
7
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Sponge nitrile coating is absorbent and provides enhanced grip in wet or oily applications.

  • Nitrile foam coating is ideal in applications that require fit, feel, and touch sensitivity
  • Gloves are absorbent and enhance grip in wet or oily applications
  • Sanitized with antimicrobial bacteriostatic lining

Specifications

ANSI Abrasion Rating ANSI 4
Coated Area Palm
Construction Seamless Knit
Coating Material Nitrile Foam
Color Black
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size 7
For Use With (Application) Wet and oily conditions
Glove Cuff Style Knit
Series 4540
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.