Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ Zorb-IT™ Ultimate Sponge Nitrile Flat Dipped Gloves
Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
Ideal for construction, oily parts handling, mechanics, automotive assembly, maintenance, cut resistance, sheet metal handling, and glass handling.
- Sponge, nitrile-coated palm
- Cut-resistant shell made from Kevlar™ fibers spun with stretchable, breathable Lycra™
- Cut-resistant, seamless aramid fiber lining, knit wrist
- Maximum Oil Absorbency (MXOA), oily grip
- Shallow-dipped, sponge gripping
- Provide form-fitting comfort
- Yellow with gray dip coating
- Launder at 104°F (40°C) using a neutral detergent
Specifications
Specifications
| Coating Material | Sponge Nitrile |
| Color | Gray, Yellow |
| Cut Resistance Rating | ANSI A3 |
| Material | Aramid Fiber, Lycra |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| For Use With (Application) | Construction, oily parts handling, mechanics, automotive assembly, maintenance, cut resistance, sheet metal handling, glass handling, recycling, shipping and receiving |
| Glove Cuff Style | Knit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction