Learn More
SHOWA™ Zorb-IT™ Nitrile Coated Work Gloves
Catalog No. 191301301C Shop All Showa Best Glove Products
Click to view available options
Size:
8
10
This item is not returnable. View return policy
This item is not returnable. View return policy
Gloves are resistant to deformation and help protect hands from oils, hydrocarbons, grease, and abrasion.
  • Sponge nitrile technology produces an extra-absorbent nitrile dip
  • Gloves absorb almost twice as much oil and grease, while still delivering a sure grip
  • Resistant to deformation
Specifications
Coated Area Palm
Construction Seamless Knit
Coating Material Nitrile
Color Gray, White
Exterior Finish No Texture
Hand Specific
Material Nylon, Polyester
Product Type Seamless Knit Gloves
Size 8
EN388 Standard 4131
Glove Cuff Style Knit
Series 4550
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.