Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP-1 Antibody (CPTC-PTPN6-2), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB383742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383742 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383742 Supplier Novus Biologicals Supplier No. NBP308900AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHP-1 Monoclonal antibody specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen SHP-1
Applications Western Blot, ELISA
Classification Monoclonal
Clone CPTC-PTPN6-2
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1
Host Species Mouse
Immunogen Recombinant human full-length SHP-1 protein (Uniprot: P29350)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5777
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.