Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP-1 Mouse anti-Human, Clone: CPTC-PTPN6-2, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP30715820UG

 View more versions of this product

Catalog No. NB121790


Only null left
Add to Cart

Description

Description

SHP-1 Monoclonal specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SHP-1
Monoclonal
0.2 mg/ml
Western Blot 1-2 ug/ml, ELISA
HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1
Recombinant human full-length SHP-1 protein
20 μg
Apoptosis, Phospho Specific, Protein Phosphatase
5777
Store at 4°C.
IgG2a κ
Western Blot, ELISA
CPTC-PTPN6-2
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.