Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHP-1 Mouse anti-Human, Clone: CPTC-PTPN6-2, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | SHP-1 |
---|---|
Clone | CPTC-PTPN6-2 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA |
Applications | Western Blot, ELISA |
Description
SHP-1 Monoclonal specifically detects SHP-1 in Human samples. It is validated for Western Blot, ELISA.Specifications
SHP-1 | |
0.2 mg/ml | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Apoptosis, Phospho Specific, Protein Phosphatase | |
10 mM PBS with 0.05% BSA | |
5777 | |
IgG2a κ | |
Protein A or G purified |
CPTC-PTPN6-2 | |
Western Blot 1-2 ug/ml, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
HCP, HCPH, HPTP1C, PTP-1C, PTPN6, SHP1, SHP-1L, SH-PTP1 | |
Recombinant human full-length SHP-1 protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title