Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP/NR0B2/Nuclear Receptor SHP Antibody, Novus Biologicals™
SDP

Catalog No. NBP15281620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15281620 20 μL
NBP152816 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15281620 Supplier Novus Biologicals Supplier No. NBP15281620UL

Rabbit Polyclonal Antibody

SHP/NR0B2/Nuclear Receptor SHP Polyclonal specifically detects SHP/NR0B2/Nuclear Receptor SHP in Human samples. It is validated for Western Blot.

Specifications

Antigen SHP/NR0B2/Nuclear Receptor SHP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q15466
Gene Alias FLJ17090, nuclear receptor subfamily 0 group B member 2, nuclear receptor subfamily 0, group B, member 2, Orphan nuclear receptor SHP, SHPSHP1, Small heterodimer partner
Gene Symbols NR0B2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NR0B2(nuclear receptor subfamily 0, group B, member 2) The peptide sequence was selected from the N terminal of NR0B2. Peptide sequence STSQPGACPCQGAASRPAILYALLSSSLKAVPRPRSRCLCRQHRPVQLCA.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 8431
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.