Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHP/NR0B2/Nuclear Receptor SHP Antibody (OTI7B5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274172A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274172A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274172A30 Supplier Novus Biologicals Supplier No. NBP274172AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SHP/NR0B2/Nuclear Receptor SHP Monoclonal specifically detects SHP/NR0B2/Nuclear Receptor SHP in Human samples. It is validated for Western Blot.

Specifications

Antigen SHP/NR0B2/Nuclear Receptor SHP
Applications Western Blot
Classification Monoclonal
Clone OTI7B5
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ17090, nuclear receptor subfamily 0 group B member 2, nuclear receptor subfamily 0, group B, member 2, Orphan nuclear receptor SHP, SHPSHP1, Small heterodimer partner
Gene Symbols NR0B2
Host Species Mouse
Immunogen Full length recombinant protein of human SHP/NR0B2/Nuclear Receptor SHP (NP_068804) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8431
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.