Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHQ1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342373 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342373 Supplier Novus Biologicals Supplier No. NBP281793MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHQ1 Polyclonal antibody specifically detects SHQ1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen SHQ1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZp686H07226, FLJ10539, protein SHQ1 homolog, SHQ1 homolog (S. cerevisiae), Shq1p
Host Species Rabbit
Immunogen SHQ1 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human SHQ1. The immunogen is located within amino acids 80 - 130 of SHQ1. Amino Acid Squence: LAPRKSRTAKPLVEEIG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55164
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.