Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shrew-1/AJAP1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB342345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342345 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342345 Supplier Novus Biologicals Supplier No. NBP281800MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Shrew-1/AJAP1 Polyclonal antibody specifically detects Shrew-1/AJAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry

Specifications

Antigen Shrew-1/AJAP1
Applications Western Blot, ELISA, Immunocytochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias adherens junction associated protein 1, adherens junction-associated protein 1, adherens junctions associated protein 1, membrane protein shrew-1, MOT8, RP3-426F10.1, SHREW1, SHREW-1, transmembrane protein SHREW1
Host Species Rabbit
Immunogen AJAP1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human AJAP1. The immunogen is located within amino acids 290 - 340 of AJAP1. Amino Acid Squence: AQSGNTRRNSHQRKTNQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55966
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.