Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHROOM3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017194 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017194 Supplier Novus Biologicals Supplier No. NBP281883
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SHROOM3 Polyclonal specifically detects SHROOM3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen SHROOM3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/mL, ELISA
Formulation PBS
Gene Alias APXL3, MSTP013, SHRM, ShrmL, SHROOM3 shroom family member 3
Gene Symbols SHROOM3
Host Species Rabbit
Immunogen Rabbit polyclonal SHROOM3 antibody was raised against an 18 amino acid peptide near the center of human SHROOM3. The immunogen is located within amino acids 850 - 900 of SHROOM3. Amino Acid Squence: CQLSGGASDSGRGPQRPDA
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57619
Test Specificity SHROOM3 antibody is human specific. At least four isoforms of SHROOM3 are known to exist; this antibody will detect all four isoforms. SHROOM3 antibody is predicted to not cross-react with other SHROOM family members.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.