Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SI-2 hydrochloride, >98%
GREENER_CHOICESDP

Catalog No. 596450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-645-0 50 mg
59-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 59-645-0 Supplier Tocris Bioscience™ Supplier No. 5964/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Steroid receptor coactivator 3 (SRC-3) inhibitor

Potent steroid receptor coactivator 3 (SRC-3) inhibitor. Reduces SRC1, SRC2 and SRC3 levels in breast cancer cell lines. Inhibits cell growth (IC50 = 3.4 nM), induces apoptosis and attenuates migration of MDA-MB48 breast cancer cells in vitro. Inhibits tumor growth in a MDA-MB48 cell breast cancer mouse model. Orally available.

Chemical Identifiers

CAS 287403-39-8
Molecular Formula C17H22N2O5S2

Specifications

Quantity 50 mg
Solubility Soluble to 50mM in water and to 20mM in DMSO
Formula Weight 301.78
Purity >98%
Molecular Weight (g/mol) 398.492
Chemical Name or Material (3S)-4-[[4-(2-Butyn-1-yloxy)phenyl]sulfonyl]-N-hydroxy-2,2-dimethyl-3-thiomorpholinecarboxamide
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.