Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIAE Antibody (03), PE, Novus Biologicals™
SDP

Catalog No. NB353333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353333 Supplier Novus Biologicals Supplier No. NBP306150PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIAE Monoclonal antibody specifically detects SIAE in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SIAE
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 03
Conjugate PE
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias AIS6, CSEC, CSE-C, cytosolic sialic acid 9-O-acetylesterase homolog, EC 3.1.1.53, H-Lse, LSE, MGC87009, sialate O-acetylesterase, sialic acid acetylesterase, Sialic acid-specific 9-O-acetylesterase, sialic acid-specific acetylesterase II, YSG2, Ysg2 homolog (mouse)
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SIAE (Uniprot#: Q9HAT2-1; Met1-Lys523).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54414
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.