Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIAH1/2 Antibody (8G7H12), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB11093596
Description
SIAH1/2 Monoclonal specifically detects SIAH1/2 in Human, Rat, Porcine, Drosophila, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot, Immunoprecipitation.Specifications
SIAH1/2 | |
Monoclonal | |
Unconjugated | |
Tissue culture supernatant with 0.1% Sodium Azide | |
E3 ubiquitin-protein ligase SIAH1, EC 6.3.2, EC 6.3.2.-, FLJ08065, hSIAH1, HUMSIAH, Seven in absentia homolog 1, seven in absentia homolog 1 (Drosophila), Siah-1, siah-1a, SIAH1A | |
Mouse | |
37 kDa | |
0.5 mL | |
Cancer, Cell Biology, Neurodegeneration, Neuroscience, Ubiquitin Proteasome Pathway | |
6477 | |
Human, Rat, Pig, Drosophila, Zebrafish, Mouse (Negative) | |
Supernatant |
Western Blot, Immunoprecipitation | |
8G7H12 | |
Western Blot 1:500, Immunoprecipitation 1:10-1:500 | |
NP_005058.2 | |
SIAH1 | |
Synthetic peptide made to a C-terminal portion of the drosophila SINA protein (within residues 280-331). [Swiss-Prot# P21461] | |
Tissue culture supernatant | |
RUO | |
Primary | |
This is specific for human SIAH 1 and 2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells. | |
Store at 4C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction