Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIAH1/2 Antibody (8G7H12), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$181.90 - $402.00

Specifications

Antigen SIAH1/2
Clone 8G7H12
Dilution Western Blot 1:500, Immunoprecipitation 1:10-1:500
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11093596T
SDP
View Documents
Novus Biologicals
NB11093596SS
0.125 mL
Each for $181.90
Only null left
Add to Cart
 
NB11093596
SDP
View Documents
Novus Biologicals
NB11093596
0.5 mL
Each for $402.00
Only null left
Add to Cart
 
Description

Description

SIAH1/2 Monoclonal specifically detects SIAH1/2 in Human, Rat, Porcine, Drosophila, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

SIAH1/2
Western Blot 1:500, Immunoprecipitation 1:10-1:500
Monoclonal
Supernatant
RUO
Human, Rat, Pig, Drosophila, Zebrafish, Mouse (Negative)
NP_005058.2
6477
Synthetic peptide made to a C-terminal portion of the drosophila SINA protein (within residues 280-331). [Swiss-Prot# P21461]
Primary
Store at 4C. Do not freeze.
37 kDa
8G7H12
Western Blot, Immunoprecipitation
Unconjugated
Mouse
Cancer, Cell Biology, Neurodegeneration, Neuroscience, Ubiquitin Proteasome Pathway
Tissue culture supernatant with 0.1% Sodium Azide
E3 ubiquitin-protein ligase SIAH1, EC 6.3.2, EC 6.3.2.-, FLJ08065, hSIAH1, HUMSIAH, Seven in absentia homolog 1, seven in absentia homolog 1 (Drosophila), Siah-1, siah-1a, SIAH1A
SIAH1
IgG1
Tissue culture supernatant
This is specific for human SIAH 1 and 2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.