Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIAH1/2 Antibody (8G7H12), Novus Biologicals™
SDP

Catalog No. NB11093596T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11093596T 0.125 mL
NB11093596 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11093596T Supplier Novus Biologicals Supplier No. NB11093596SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

SIAH1/2 Monoclonal specifically detects SIAH1/2 in Human, Rat, Porcine, Drosophila, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen SIAH1/2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 8G7H12
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunoprecipitation 1:10-1:500
Formulation Tissue culture supernatant with 0.1% Sodium Azide
Gene Accession No. NP_005058.2
Gene Alias E3 ubiquitin-protein ligase SIAH1, EC 6.3.2, EC 6.3.2.-, FLJ08065, hSIAH1, HUMSIAH, Seven in absentia homolog 1, seven in absentia homolog 1 (Drosophila), Siah-1, siah-1a, SIAH1A
Gene Symbols SIAH1
Host Species Mouse
Immunogen Synthetic peptide made to a C-terminal portion of the drosophila SINA protein (within residues 280-331). [Swiss-Prot# P21461]
Molecular Weight of Antigen 37 kDa
Purification Method Tissue culture supernatant
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Neurodegeneration, Neuroscience, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6477
Test Specificity This is specific for human SIAH 1 and 2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells.
Target Species Human, Rat, Pig, Drosophila, Zebrafish, Mouse (Negative)
Content And Storage Store at 4C. Do not freeze.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.